Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester dicyclohexylammonium salt, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester dicyclohexylammonium salt
CAS 34897-61-5
Formula Weight 518.69
MDL Number MFCD00077017
Beilstein 4286980
Synonym Z-Glu-OtBu.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6656403

 
alfa aesar™
H6656403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6656406

 
alfa aesar™
H6656406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 518.69
Physical Form Solid
CAS 34897-61-5
MDL Number MFCD00077017
Assay 95%
Storage Note 1 -30° to -10°C
Quantity 1g
Synonym Z-Glu-OtBu.DCHA
Beilstein 4286980
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester dicyclohexylammonium salt
Safety and Handling

-30° to -10°C