Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-aspartic acid, 98+%

$29.99 - $103.07

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid
CAS 1152-61-0
Molecular Formula C12H13NO6
Formula Weight 267.24
MDL Number MFCD00002719
Beilstein 2336722
Synonym N-Cbz-L-aspartic acid; Z-Asp-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0874514

  SDS
alfa aesar™
L0874514
25g Each for $103.07

AAL0874506

  SDS
alfa aesar™
L0874506
5g Each for $29.99
Description & Specifications

Specifications

Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid
CAS 1152-61-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
TSCA TSCA
Assay ≥98%
Molecular Formula C12H13NO6
Formula Weight 267.24
Melting Point 116° to 121°C
MDL Number MFCD00002719
Beilstein 2336722
Quantity 25g
Synonym N-Cbz-L-aspartic acid; Z-Asp-OH
Optical Rotation +9° (c=2 in Acetic Acid)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C