missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester, 95%

$40.72 - $532.03

Chemical Identifiers

CAS 3338-32-7
Molecular Formula C20H24N2O8
Molecular Weight (g/mol) 420.418
MDL Number MFCD00153328
InChI Key JUMSBOKRGDETHL-AWEZNQCLSA-N
Synonym z-asp otbu-osu, 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-benzyloxy carbonyl amino butanedioate, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate, z-asp obut-osu, n-cbz-, a-t-butyl-l-aspartic acid n-hydroxysuccinimide ester, z-l-aspartic acid 4-tert-butyl-1-n-succinimidyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl-2-benzyloxy carbonyl amino succinate, n-alpha-benzyloxycarbonyl-l-aspartic acid alpha-n-hydroxysuccinimidyl beta-t-butyl ester, tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate
PubChem CID 11026196
IUPAC Name 4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(phenylmethoxycarbonylamino)butanedioate
SMILES CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65816MD View Documents Alfa Aesar™
H65816MD
250mg Each for $40.72
 
AAH6581603 View Documents Alfa Aesar™
H6581603
1g Each for $128.31
 
AAH6581606 View Documents Alfa Aesar™
H6581606
5g Each for $532.03
 
Specifications

Chemical Identifiers

3338-32-7
420.418
JUMSBOKRGDETHL-AWEZNQCLSA-N
11026196
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2
C20H24N2O8
MFCD00153328
z-asp otbu-osu, 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-benzyloxy carbonyl amino butanedioate, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate, z-asp obut-osu, n-cbz-, a-t-butyl-l-aspartic acid n-hydroxysuccinimide ester, z-l-aspartic acid 4-tert-butyl-1-n-succinimidyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl-2-benzyloxy carbonyl amino succinate, n-alpha-benzyloxycarbonyl-l-aspartic acid alpha-n-hydroxysuccinimidyl beta-t-butyl ester, tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(phenylmethoxycarbonylamino)butanedioate

Specifications

3338-32-7
MFCD00153328
z-asp otbu-osu, 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-benzyloxy carbonyl amino butanedioate, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate, z-asp obut-osu, n-cbz-, a-t-butyl-l-aspartic acid n-hydroxysuccinimide ester, z-l-aspartic acid 4-tert-butyl-1-n-succinimidyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl-2-benzyloxy carbonyl amino succinate, n-alpha-benzyloxycarbonyl-l-aspartic acid alpha-n-hydroxysuccinimidyl beta-t-butyl ester, tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2
420.418
420.42
95%
148°C to 152°C
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester
C20H24N2O8
1558119
JUMSBOKRGDETHL-AWEZNQCLSA-N
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(phenylmethoxycarbonylamino)butanedioate
11026196
Powder
White
250mg
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C