missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6581606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
148°C to 152°C | |
3338-32-7 | |
C20H24N2O8 | |
1558119 | |
JUMSBOKRGDETHL-AWEZNQCLSA-N | |
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(phenylmethoxycarbonylamino)butanedioate | |
11026196 | |
95% | |
Powder |
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester | |
White | |
MFCD00153328 | |
z-asp otbu-osu, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate, n-cbz-, n-cbz-, a-t-butyl-l-aspartic acid n-hydroxysuccinimide ester, n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester, n-alpha-benzyloxycarbonyl-l-aspartic acid alpha-n-hydroxysuccinimidyl beta-t-butyl ester, tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate, tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate | |
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2 | |
420.418 | |
420.42 | |
5 g |
Chemical Identifiers
3338-32-7 | |
420.418 | |
JUMSBOKRGDETHL-AWEZNQCLSA-N | |
11026196 | |
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2 |
C20H24N2O8 | |
MFCD00153328 | |
z-asp otbu-osu, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate, s-4-tert-butyl 1-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino succinate, n-cbz-, n-cbz-, a-t-butyl-l-aspartic acid n-hydroxysuccinimide ester, n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-succinimidyl 4-tert-butyl ester, n-alpha-benzyloxycarbonyl-l-aspartic acid alpha-n-hydroxysuccinimidyl beta-t-butyl ester, tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate, tert-butyl 2,5-dioxoazolidinyl 2s-2-phenylmethoxy carbonylamino butane-1,4-dioate | |
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-(phenylmethoxycarbonylamino)butanedioate |
Safety and Handling
EINECSNumber : 222-080-8
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only