Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-methyl ester, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-methyl ester
CAS 63327-57-1
Formula Weight 337.37
MDL Number MFCD00270581
Beilstein 2601725
Synonym Z-Asp(OtBu)-OMe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6638406

 
alfa aesar™
H6638406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6638414

 
alfa aesar™
H6638414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 337.37
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-methyl ester
MDL Number MFCD00270581
Assay 98%
Physical Form Powder
Synonym Z-Asp(OtBu)-OMe
Beilstein 2601725
Storage Note 1 -30° to -10°C
Quantity 5g
Color Off-white to white
CAS 63327-57-1
Safety and Handling

-30° to -10°C