missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester, 98%

Manufacturer:  Alfa Aesar™ H6364203

 View more versions of this product

Catalog No. AAH6364203



Specifications

Specifications

N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester
C13H15NO6
6279422
PHMBNDDHIBIDRQ-JTQLQIEISA-N
(2S)-4-methoxy-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
7014888
Powder
White
3160-47-2
MFCD00133585
z-asp ome-oh, s-2-n-cbz-amino-succinic acid 4-methyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-methyl ester, s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, cbz-l-aspartic acid 4-methyl ester, n-cbz-l-aspartate, pubchem14062, z-l-aspartic acid 4-methyl ester, z-asp ome-oh tlc
COC(=O)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
281.264
281.26
98%
1g

Chemical Identifiers

3160-47-2
281.264
PHMBNDDHIBIDRQ-JTQLQIEISA-N
7014888
COC(=O)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C13H15NO6
MFCD00133585
z-asp ome-oh, s-2-n-cbz-amino-succinic acid 4-methyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-methyl ester, s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, cbz-l-aspartic acid 4-methyl ester, n-cbz-l-aspartate, pubchem14062, z-l-aspartic acid 4-methyl ester, z-asp ome-oh tlc
(2S)-4-methoxy-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents