Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester
CAS 3160-47-2
Formula Weight 281.26
MDL Number MFCD00133585
Beilstein 6279422
Synonym Z-Asp(OMe)-OH; (S)-2-(Benzyloxycarbonylamino)-4-methoxy-4-oxobutyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6364203

  SDS
alfa aesar™
H6364203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6364206

  SDS
alfa aesar™
H6364206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 281.26
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester
Physical Form Powder
Assay 98%
MDL Number MFCD00133585
CAS 3160-47-2
Beilstein 6279422
Synonym Z-Asp(OMe)-OH; (S)-2-(Benzyloxycarbonylamino)-4-methoxy-4-oxobutyric acid
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C