Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-alaninol

$97.74 - $255.53

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-alaninol
CAS 66674-16-6
Molecular Formula C11H15NO3
Formula Weight 209.25
MDL Number MFCD00672530
Synonym (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester; Z-Ala-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2706603

 
alfa aesar™
H2706603
1g Each for $97.74

AAH2706606

 
alfa aesar™
H2706606
5g Each for $255.53
Description & Specifications

Specifications

Formula Weight 209.25
Chemical Name or Material N-Benzyloxycarbonyl-L-alaninol
Molecular Formula C11H15NO3
MDL Number MFCD00672530
CAS 66674-16-6
Quantity 1g
Synonym (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester; Z-Ala-ol

Applications
N-Benzyloxycarbonyl-L-alaninol is used as an important organic intermediate. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.