missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-alanine methyl ester, 95%

$67.08 - $293.00

Chemical Identifiers

CAS 28819-05-8
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00270464
InChI Key OMDVFTVXPVXANK-VIFPVBQESA-N
Synonym z-ala-ome, z-l-alanine methyl ester, methyl 2s-2-benzyloxy carbonyl amino propanoate, n-trityl-l-ala, cbz-ala-ome, z-l-alaninemethylester, n-z-alanine methyl ester, n-cbz-l-alanine methyl ester, methyl n-benzyloxycarbonylalaninate, n-carbobenzyloxy-l-alanine methyl ester
PubChem CID 6993447
IUPAC Name methyl (2S)-2-(phenylmethoxycarbonylamino)propanoate
SMILES CC(C(=O)OC)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6274906 View Documents Alfa Aesar™
H6274906
5g Each for $67.08
 
AAH6274914 View Documents Alfa Aesar™
H6274914
25g Each for $293.00
 
Specifications

Chemical Identifiers

28819-05-8
237.255
OMDVFTVXPVXANK-VIFPVBQESA-N
6993447
CC(C(=O)OC)NC(=O)OCC1=CC=CC=C1
C12H15NO4
MFCD00270464
z-ala-ome, z-l-alanine methyl ester, methyl 2s-2-benzyloxy carbonyl amino propanoate, n-trityl-l-ala, cbz-ala-ome, z-l-alaninemethylester, n-z-alanine methyl ester, n-cbz-l-alanine methyl ester, methyl n-benzyloxycarbonylalaninate, n-carbobenzyloxy-l-alanine methyl ester
methyl (2S)-2-(phenylmethoxycarbonylamino)propanoate

Specifications

5g
C12H15NO4
z-ala-ome, z-l-alanine methyl ester, methyl 2s-2-benzyloxy carbonyl amino propanoate, n-trityl-l-ala, cbz-ala-ome, z-l-alaninemethylester, n-z-alanine methyl ester, n-cbz-l-alanine methyl ester, methyl n-benzyloxycarbonylalaninate, n-carbobenzyloxy-l-alanine methyl ester
CC(C(=O)OC)NC(=O)OCC1=CC=CC=C1
237.255
237.25
N-Benzyloxycarbonyl-L-alanine methyl ester
28819-05-8
MFCD00270464
OMDVFTVXPVXANK-VIFPVBQESA-N
methyl (2S)-2-(phenylmethoxycarbonylamino)propanoate
6993447
95%