missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-2-cyclohexylglycine, 95%

Manufacturer:  Alfa Aesar™ H6372303

 View more versions of this product

Catalog No. AAH6372303



Specifications

Specifications

N-Benzyloxycarbonyl-L-2-cyclohexylglycine
C16H21NO4
z-chg-oh, s-2-benzyloxy carbonyl amino-2-cyclohexylacetic acid, s-cbz-cyclohexylglycine, cbz-cyclohexyl-l-glycine, pubchem5639, 2s-benzyloxy carbonyl amino cyclohexyl acetic acid, n-benzyloxycarbonyl-2-cyclohexyl-l-glycine, s-benzyloxycarbonylamino-cyclohexyl-acetic acid, s-2-benzyloxy carbonyl amino-2-cyclohexylaceti, 2s-2-benzyloxycarbonylamino-2-cyclohexyl-acetic acid
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2=CC=CC=C2
291.347
291.35
95%
1g
69901-75-3
MFCD00191083
CUSYTUPJAYLNFQ-AWEZNQCLSA-N
(2S)-2-cyclohexyl-2-(phenylmethoxycarbonylamino)acetic acid
16218812
Solid
White

Chemical Identifiers

69901-75-3
291.347
CUSYTUPJAYLNFQ-AWEZNQCLSA-N
16218812
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2=CC=CC=C2
C16H21NO4
MFCD00191083
z-chg-oh, s-2-benzyloxy carbonyl amino-2-cyclohexylacetic acid, s-cbz-cyclohexylglycine, cbz-cyclohexyl-l-glycine, pubchem5639, 2s-benzyloxy carbonyl amino cyclohexyl acetic acid, n-benzyloxycarbonyl-2-cyclohexyl-l-glycine, s-benzyloxycarbonylamino-cyclohexyl-acetic acid, s-2-benzyloxy carbonyl amino-2-cyclohexylaceti, 2s-2-benzyloxycarbonylamino-2-cyclohexyl-acetic acid
(2S)-2-cyclohexyl-2-(phenylmethoxycarbonylamino)acetic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents