Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-L-2-cyclohexylglycine, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-L-2-cyclohexylglycine
CAS 69901-75-3
Formula Weight 291.35
MDL Number MFCD00191083
Synonym Z-L-Chg-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6372303

  SDS
alfa aesar™
H6372303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6372306

  SDS
alfa aesar™
H6372306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.35
Quantity 1g
Color White
Assay 95%
Physical Form Solid
MDL Number MFCD00191083
Storage Note 1 -30° to -10°C
CAS 69901-75-3
Chemical Name or Material N-Benzyloxycarbonyl-L-2-cyclohexylglycine
Synonym Z-L-Chg-OH
Safety and Handling

-30° to -10°C