missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Benzyloxycarbonyl)glycylglycyl-L-valine, 97%

$199.76 - $615.91

Chemical Identifiers

CAS 34152-86-8
Molecular Formula C17H23N3O6
Molecular Weight (g/mol) 365.386
MDL Number MFCD00056144
InChI Key OLTJNTLCZYWHKR-OAHLLOKOSA-N
Synonym 2s-2-2-2-benzyloxy carbonyl amino acetamido acetamido-3-methylbutanoic acid, 2s-3-methyl-2-2-2-phenylmethoxycarbonylamino acetyl amino acetyl amino butanoic acid, n-benzyloxycarbonylglycylglycyl-l-valine, n-cbz-gly-gly-val, z-gly-gly-val-oh
PubChem CID 7079158
IUPAC Name (2R)-3-methyl-2-[[2-[[2-(phenylmethoxycarbonylamino)acetyl]amino]acetyl]amino]butanoic acid
SMILES CC(C)C(C(=O)O)NC(=O)CNC(=O)CNC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65992MD View Documents Alfa Aesar™
H65992MD
250mg Each for $199.76
 
AAH6599203 View Documents Alfa Aesar™
H6599203
1g Each for $615.91
 
Specifications

Chemical Identifiers

34152-86-8
365.386
OLTJNTLCZYWHKR-OAHLLOKOSA-N
7079158
CC(C)C(C(=O)O)NC(=O)CNC(=O)CNC(=O)OCC1=CC=CC=C1
C17H23N3O6
MFCD00056144
2s-2-2-2-benzyloxy carbonyl amino acetamido acetamido-3-methylbutanoic acid, 2s-3-methyl-2-2-2-phenylmethoxycarbonylamino acetyl amino acetyl amino butanoic acid, n-benzyloxycarbonylglycylglycyl-l-valine, n-cbz-gly-gly-val, z-gly-gly-val-oh
(2R)-3-methyl-2-[[2-[[2-(phenylmethoxycarbonylamino)acetyl]amino]acetyl]amino]butanoic acid

Specifications

34152-86-8
MFCD00056144
2s-2-2-2-benzyloxy carbonyl amino acetamido acetamido-3-methylbutanoic acid, 2s-3-methyl-2-2-2-phenylmethoxycarbonylamino acetyl amino acetyl amino butanoic acid, n-benzyloxycarbonylglycylglycyl-l-valine, n-cbz-gly-gly-val, z-gly-gly-val-oh
CC(C)C(C(=O)O)NC(=O)CNC(=O)CNC(=O)OCC1=CC=CC=C1
365.386
365.39
97%
N-(Benzyloxycarbonyl)glycylglycyl-L-valine
C17H23N3O6
2490015
OLTJNTLCZYWHKR-OAHLLOKOSA-N
(2R)-3-methyl-2-[[2-[[2-(phenylmethoxycarbonylamino)acetyl]amino]acetyl]amino]butanoic acid
7079158
Solid
250mg
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures