Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-DL-valine, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-DL-valine
CAS 3588-63-4
Formula Weight 251.28
MDL Number MFCD00065127
Beilstein 2056615
Synonym Z-DL-Val-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6678306

 
alfa aesar™
H6678306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6678314

 
alfa aesar™
H6678314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.28
CAS 3588-63-4
MDL Number MFCD00065127
Assay 98%
Synonym Z-DL-Val-OH
Physical Form Powder
Storage Note 1 2° to 8°C
Color Off-white to white
TSCA TSCA
Beilstein 2056615
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-DL-valine
Safety and Handling

2° to 8°C