Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-DL-proline, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-DL-proline
CAS 5618-96-2
Formula Weight 249.27g/mol
MDL Number MFCD00063229
Synonym Z-DL-Pro-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6615503

 
alfa aesar™
H66155-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6615506

 
alfa aesar™
H66155-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Color Off-white to white
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonyl-DL-proline
CAS 5618-96-2
Physical Form Powder
Assay 98%
Formula Weight 249.27g/mol
MDL Number MFCD00063229
Synonym Z-DL-Pro-OH
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C