Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-DL-norvaline, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-DL-norvaline
CAS 21691-43-0
Formula Weight 251.28
MDL Number MFCD00020434
Beilstein 3209289
Synonym Z-DL-Nva-OH; 2-(Benzyloxycarbonylamino)pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6649806

 
alfa aesar™
H6649806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6649814

 
alfa aesar™
H6649814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.28
Chemical Name or Material N-Benzyloxycarbonyl-DL-norvaline
Assay 98%
MDL Number MFCD00020434
Synonym Z-DL-Nva-OH; 2-(Benzyloxycarbonylamino)pentanoic acid
Physical Form Powder
CAS 21691-43-0
Beilstein 3209289
Color Off-white to white
Quantity 5g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C