Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-DL-aspartic acid, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-DL-aspartic acid
CAS 4515-21-3
Formula Weight 267.24
MDL Number MFCD00063181
Beilstein 4701629
Synonym Z-DL-Asp-OH; N-Cbz-DL-aspartic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6695006

 
alfa aesar™
H6695006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6695014

 
alfa aesar™
H6695014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6695022

 
alfa aesar™
H6695022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.24
Chemical Name or Material N-Benzyloxycarbonyl-DL-aspartic acid
MDL Number MFCD00063181
Assay 95%
Physical Form Powder
Synonym Z-DL-Asp-OH; N-Cbz-DL-aspartic acid
Storage Note 1 -30° to -10°C
Beilstein 4701629
Color White
Quantity 5g
CAS 4515-21-3
Safety and Handling

-30° to -10°C