Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-DL-asparagine, 98%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-DL-asparagine
CAS 29880-22-6
Formula Weight 266.25
MDL Number MFCD00065101
Beilstein 6066596
Synonym Z-DL-Asn-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6658706

 
alfa aesar™
H6658706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6658714

 
alfa aesar™
H6658714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6658722

 
alfa aesar™
H6658722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 266.25
CAS 29880-22-6
MDL Number MFCD00065101
Assay 98%
Chemical Name or Material N-Benzyloxycarbonyl-DL-asparagine
Beilstein 6066596
Synonym Z-DL-Asn-OH
Physical Form Powder
Color White
Quantity 5g