missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-D-threonine, 98+%, Thermo Scientific™
$35.45 - $35.45
Chemical Identifiers
CAS | 80384-27-6 |
---|---|
Molecular Formula | C12H15NO5 |
Molecular Weight (g/mol) | 253.25 |
MDL Number | MFCD00037808 |
InChI Key | IPJUIRDNBFZGQN-WCBMZHEXSA-N |
Synonym | z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh |
PubChem CID | 853484 |
IUPAC Name | (2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid |
SMILES | C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL08561MD
|
Thermo Scientific Chemicals
L08561MD |
250 mg |
Each for $35.45
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
80384-27-6 | |
253.25 | |
IPJUIRDNBFZGQN-WCBMZHEXSA-N | |
853484 | |
C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
C12H15NO5 | |
MFCD00037808 | |
z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh | |
(2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid |
Specifications
80384-27-6 | |
C12H15NO5 | |
250 mg | |
z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh | |
+5.5° (c=2 in Acetic Acid) | |
(2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid | |
853484 | |
≥98% |
100°C to 105°C | |
MFCD00037808 | |
4695777 | |
IPJUIRDNBFZGQN-WCBMZHEXSA-N | |
C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O | |
253.25 | |
253.26 | |
N-Benzyloxycarbonyl-D-threonine |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold