Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-serine, 98+%

$39.93 - $39.93

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-serine
CAS 6081-61-4
Molecular Formula C11H13NO5
Formula Weight 239.23
MDL Number MFCD00063144
Beilstein 2058313
Synonym N-Cbz-D-serine; Z-D-Ser-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0838403

  SDS
alfa aesar™
L0838403
1g Each for $39.93

AAL0838406

  SDS
alfa aesar™
L0838406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.23
Chemical Name or Material N-Benzyloxycarbonyl-D-serine
CAS 6081-61-4
Molecular Formula C11H13NO5
MDL Number MFCD00063144
Assay ≥98%
Synonym N-Cbz-D-serine; Z-D-Ser-OH
Beilstein 2058313
Quantity 1g
Melting Point 116° to 119°C
Optical Rotation −6° (c=7 in Acetic Acid)