Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-proline, 98+%

$32.54 - $66.45

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-proline
CAS 6404-31-5
Molecular Formula C13H15NO4
Formula Weight 249.27
MDL Number MFCD00063228
Beilstein 485188
Synonym N-Cbz-D-proline; Z-D-Pro-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2500503

  SDS
alfa aesar™
B2500503
1g Each for $32.54

AAB2500506

  SDS
alfa aesar™
B2500506
5g Each for $66.45

AAB2500514

  SDS
alfa aesar™
B2500514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.27
Chemical Name or Material N-Benzyloxycarbonyl-D-proline
Assay ≥98%
Molecular Formula C13H15NO4
Melting Point 76° to 78°C
MDL Number MFCD00063228
CAS 6404-31-5
Beilstein 485188
Quantity 1g
Synonym N-Cbz-D-proline; Z-D-Pro-OH
Optical Rotation +40° (c=2 in Ethanol)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C