missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-D-proline, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2500506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Benzyloxycarbonyl-D-proline | |
76°C to 78°C | |
MFCD00063228 | |
485188 | |
JXGVXCZADZNAMJ-LLVKDONJSA-N | |
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O | |
249.266 | |
249.27 |
6404-31-5 | |
C13H15NO4 | |
5 g | |
n-benzyloxycarbonyl-d-proline, z-d-pro-oh, +-z-d-proline, z-d-proline, r-n-benzyloxycarbonylproline, n-cbz-d-proline, n-carbobenzoxy-d-proline, 1-benzyloxy carbonyl-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid | |
+40° (c=2 in Ethanol) | |
(2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid | |
111208 | |
≥98% |
Chemical Identifiers
6404-31-5 | |
249.266 | |
JXGVXCZADZNAMJ-LLVKDONJSA-N | |
111208 | |
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
C13H15NO4 | |
MFCD00063228 | |
n-benzyloxycarbonyl-d-proline, z-d-pro-oh, +-z-d-proline, z-d-proline, r-n-benzyloxycarbonylproline, n-cbz-d-proline, n-carbobenzoxy-d-proline, 1-benzyloxy carbonyl-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid | |
(2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid |
Safety and Handling
EINECSNumber : 229-021-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only