missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyloxycarbonyl-D-phenylalaninol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2781906

 View more versions of this product

Catalog No. AAH2781906



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

92°C to 95°C
58917-85-4
MFCD00191193
WPOFMMJJCPZPAO-MRXNPFEDSA-N
benzyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
736170
98%
+42° (c=1 in Methanol)
N-Benzyloxycarbonyl-D-phenylalaninol
C17H19NO3
cbz-d-phenylalaninol, z-d-phenylalaninol, n-carbobenzoxy-d-phenylalaninol, r-+-2-cbz-amino-3-phenyl-1-propanol, r-+-2-benzyloxycarbonylamino-3-phenyl-1-propanol, n-carbobenzyloxy-d-phenylalaninol, r-+-2-carbobenzyloxyamino-3-phenyl-1-propanol, benzyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate, r-2-z-amino-3-phenyl-1-propanol, benzyl 2r-1-hydroxy-3-phenylpropan-2-yl carbamate
OC[C@@H](CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
285.34
285.35
5 g

Chemical Identifiers

58917-85-4
285.34
WPOFMMJJCPZPAO-MRXNPFEDSA-N
736170
OC[C@@H](CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
C17H19NO3
MFCD00191193
cbz-d-phenylalaninol, z-d-phenylalaninol, n-carbobenzoxy-d-phenylalaninol, r-+-2-cbz-amino-3-phenyl-1-propanol, r-+-2-benzyloxycarbonylamino-3-phenyl-1-propanol, n-carbobenzyloxy-d-phenylalaninol, r-+-2-carbobenzyloxyamino-3-phenyl-1-propanol, benzyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate, r-2-z-amino-3-phenyl-1-propanol, benzyl 2r-1-hydroxy-3-phenylpropan-2-yl carbamate
benzyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only