missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-D-phenylalanine, 98+%, Thermo Scientific™
$40.45 - $172.16
Chemical Identifiers
CAS | 2448-45-5 |
---|---|
Molecular Formula | C17H17NO4 |
Molecular Weight (g/mol) | 299.326 |
MDL Number | MFCD00063151 |
InChI Key | RRONHWAVOYADJL-OAHLLOKOSA-N |
Synonym | z-d-phe-oh, n-cbz-d-phenylalanine, z-d-phenylalanine, cbz-d-phe-oh, n-carbobenzyloxy-d-phenylalanine, chembl63865, benzyloxycarbonyl-d-phenylalanine, n-benzyloxycarbonyl-d-phenylalanine, 2r-3-phenyl-2-phenylmethoxycarbonylamino propanoic acid, d-phenylalanine, n-phenylmethoxy carbonyl |
PubChem CID | 675678 |
IUPAC Name | (2R)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid |
SMILES | C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0910203
|
Thermo Scientific Chemicals
L0910203 |
1 g |
Each for $40.45
|
|
|||||
AAL0910206
|
Thermo Scientific Chemicals
L0910206 |
5 g |
Each for $172.16
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsN-Benzyloxycarbonyl-D-phenylalanine is used for affinity chromatography of proteinases such as the extracellular fibrinogenolytic enzyme of Aspergillus fumigatus. It serves as a primary precursor for the biosynthesis of catecholamines in the body. Also used to antagonize stress-induced analgesia in humans, and also acts as an anti-enkephalinase agent.
Solubility
Soluble in methanol. Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
2448-45-5 | |
299.326 | |
RRONHWAVOYADJL-OAHLLOKOSA-N | |
675678 | |
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2 |
C17H17NO4 | |
MFCD00063151 | |
z-d-phe-oh, n-cbz-d-phenylalanine, z-d-phenylalanine, cbz-d-phe-oh, n-carbobenzyloxy-d-phenylalanine, chembl63865, benzyloxycarbonyl-d-phenylalanine, n-benzyloxycarbonyl-d-phenylalanine, 2r-3-phenyl-2-phenylmethoxycarbonylamino propanoic acid, d-phenylalanine, n-phenylmethoxy carbonyl | |
(2R)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
2448-45-5 | |
C17H17NO4 | |
1 g | |
z-d-phe-oh, n-cbz-d-phenylalanine, z-d-phenylalanine, cbz-d-phe-oh, n-carbobenzyloxy-d-phenylalanine, chembl63865, benzyloxycarbonyl-d-phenylalanine, n-benzyloxycarbonyl-d-phenylalanine, 2r-3-phenyl-2-phenylmethoxycarbonylamino propanoic acid, d-phenylalanine, n-phenylmethoxy carbonyl | |
RRONHWAVOYADJL-OAHLLOKOSA-N | |
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2 | |
299.326 | |
299.33 | |
N-Benzyloxycarbonyl-D-phenylalanine |
85°C to 87°C | |
MFCD00063151 | |
2817463 | |
Soluble in methanol. Slightly soluble in water. | |
−5° (c=5 in Acetic Acid) | |
(2R)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid | |
675678 | |
≥98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures