missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-D-phenylalaninamide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6597203
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Benzyloxycarbonyl-D-phenylalaninamide | |
C17H18N2O3 | |
1 g | |
HHBOFAIEPRHUSR-UHFFFAOYNA-N | |
benzyl N-(1-carbamoyl-2-phenylethyl)carbamate | |
68077307 | |
97% |
5241-56-5 | |
MFCD00191136 | |
r-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate, z-d-phe-nh2, benzyl n-1r-1-carbamoyl-2-phenylethyl carbamate, nalpha-benzyloxy carbonyl-d-phenylalaninamide, r-benzyl 1-amino-1-oxo-3-phenylpropan-2-ylcarbamate | |
NC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1 | |
298.34 | |
298.34 | |
Solid |
Chemical Identifiers
5241-56-5 | |
298.34 | |
HHBOFAIEPRHUSR-UHFFFAOYNA-N | |
68077307 | |
NC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1 |
C17H18N2O3 | |
MFCD00191136 | |
r-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate, z-d-phe-nh2, benzyl n-1r-1-carbamoyl-2-phenylethyl carbamate, nalpha-benzyloxy carbonyl-d-phenylalaninamide, r-benzyl 1-amino-1-oxo-3-phenylpropan-2-ylcarbamate | |
benzyl N-(1-carbamoyl-2-phenylethyl)carbamate |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold