missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-phenylalaninamide, 97%

$68.31 - $282.20

Chemical Identifiers

CAS 5241-56-5
Molecular Formula C17H18N2O3
Molecular Weight (g/mol) 298.342
MDL Number MFCD00191136
InChI Key HHBOFAIEPRHUSR-OAHLLOKOSA-N
Synonym r-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate, z-d-phe-nh2, benzyl n-1r-1-carbamoyl-2-phenylethyl carbamate, nalpha-benzyloxy carbonyl-d-phenylalaninamide, r-benzyl 1-amino-1-oxo-3-phenylpropan-2-ylcarbamate
PubChem CID 68077307
IUPAC Name benzyl N-[(2R)-1-amino-1-oxo-3-phenylpropan-2-yl]carbamate
SMILES C1=CC=C(C=C1)CC(C(=O)N)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6597203 View Documents Alfa Aesar™
H6597203
1g Each for $68.31
 
AAH6597206 View Documents Alfa Aesar™
H6597206
5g Each for $282.20
 
Specifications

Chemical Identifiers

5241-56-5
298.342
HHBOFAIEPRHUSR-OAHLLOKOSA-N
68077307
C1=CC=C(C=C1)CC(C(=O)N)NC(=O)OCC2=CC=CC=C2
C17H18N2O3
MFCD00191136
r-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate, z-d-phe-nh2, benzyl n-1r-1-carbamoyl-2-phenylethyl carbamate, nalpha-benzyloxy carbonyl-d-phenylalaninamide, r-benzyl 1-amino-1-oxo-3-phenylpropan-2-ylcarbamate
benzyl N-[(2R)-1-amino-1-oxo-3-phenylpropan-2-yl]carbamate

Specifications

1g
C17H18N2O3
r-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate, z-d-phe-nh2, benzyl n-1r-1-carbamoyl-2-phenylethyl carbamate, nalpha-benzyloxy carbonyl-d-phenylalaninamide, r-benzyl 1-amino-1-oxo-3-phenylpropan-2-ylcarbamate
C1=CC=C(C=C1)CC(C(=O)N)NC(=O)OCC2=CC=CC=C2
298.342
298.34
97%
5241-56-5
MFCD00191136
HHBOFAIEPRHUSR-OAHLLOKOSA-N
benzyl N-[(2R)-1-amino-1-oxo-3-phenylpropan-2-yl]carbamate
68077307
Solid
N-Benzyloxycarbonyl-D-phenylalaninamide
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C