Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-norvaline, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-norvaline
CAS 42918-89-8
Formula Weight 251.48
MDL Number MFCD00136826
Beilstein 2466156
Synonym Z-D-Nva-OH; (R)-2-(Benzyloxycarbonylamino)pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6636806

 
alfa aesar™
H6636806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6636814

 
alfa aesar™
H6636814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.48
Melting Point 84° to 90°C
Chemical Name or Material N-Benzyloxycarbonyl-D-norvaline
CAS 42918-89-8
Physical Form Powder
Assay 95%
Synonym Z-D-Nva-OH; (R)-2-(Benzyloxycarbonylamino)pentanoic acid
Quantity 5g
Beilstein 2466156
Storage Note 1 -30° to -10°C
Color White
MDL Number MFCD00136826
Safety and Handling

-30° to -10°C