Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-norleucine, 98+%

$54.65 - $145.31

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-norleucine
CAS 15027-14-2
Molecular Formula C14H19NO4
Formula Weight 265.3
MDL Number MFCD00063169
Beilstein 5752565
Synonym N-Cbz-D-norleucine; Z-D-Nle-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL08921MD

  SDS
alfa aesar™
L08921MD
250mg Each for $54.65

AAL0892103

  SDS
alfa aesar™
L0892103
1g Each for $145.31
Description & Specifications

Specifications

Formula Weight 265.3
Chemical Name or Material N-Benzyloxycarbonyl-D-norleucine
MDL Number MFCD00063169
CAS 15027-14-2
Molecular Formula C14H19NO4
Melting Point 56° to 58°C
Beilstein 5752565
Quantity 250mg
Synonym N-Cbz-D-norleucine; Z-D-Nle-OH
Optical Rotation +10° (c=1 in Methanol)
Assay ≥98%