Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-leucine, 95%

$30.01 - $30.01

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-leucine
CAS 28862-79-5
Molecular Formula C14H19NO4
Formula Weight 265.3
MDL Number MFCD00066068
Beilstein 2057659
Synonym N-Cbz-D-leucine; Z-D-Leu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0839703

 
alfa aesar™
L0839703
1g Each for $30.01
Description & Specifications

Specifications

Formula Weight 265.3
Chemical Name or Material N-Benzyloxycarbonyl-D-leucine
Assay 95%
CAS 28862-79-5
Molecular Formula C14H19NO4
MDL Number MFCD00066068
Beilstein 2057659
Quantity 1g
Synonym N-Cbz-D-leucine; Z-D-Leu-OH
Optical Rotation +16° (c=2 in Ethanol)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C