missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-glutamic acid, 95%

CAS: 63648-73-7 | C13H15NO6 | 281.264 g/mol

$192.42 - $603.74

Chemical Identifiers

CAS 63648-73-7
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00063193
InChI Key PVFCXMDXBIEMQG-SNVBAGLBSA-N
Synonym z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu
PubChem CID 1607568
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6140114
View Documents
Thermo Scientific Chemicals
H6140114
25 g
Each for $192.42
Only null left
 
AAH6140122
View Documents
Thermo Scientific Chemicals
H6140122
100 g
Each for $603.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63648-73-7
281.264
PVFCXMDXBIEMQG-SNVBAGLBSA-N
1607568
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O
C13H15NO6
MFCD00063193
z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu
(2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid

Specifications

63648-73-7
C13H15NO6
25 g
PVFCXMDXBIEMQG-SNVBAGLBSA-N
(2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid
1607568
95%
119°C to 121°C
MFCD00063193
z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O
281.264
281.26
N-Benzyloxycarbonyl-D-glutamic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only