missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-D-glutamic acid, 95%
CAS: 63648-73-7 | C13H15NO6 | 281.264 g/mol
Supplier: Thermo Scientific Chemicals H6140114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Benzyloxycarbonyl-D-glutamic acid | |
| 119°C to 121°C | |
| MFCD00063193 | |
| z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu | |
| C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O | |
| 281.264 | |
| 281.26 |
| 63648-73-7 | |
| C13H15NO6 | |
| 25 g | |
| PVFCXMDXBIEMQG-SNVBAGLBSA-N | |
| (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid | |
| 1607568 | |
| 95% |
Chemical Identifiers
| 63648-73-7 | |
| 281.264 | |
| PVFCXMDXBIEMQG-SNVBAGLBSA-N | |
| 1607568 | |
| C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O |
| C13H15NO6 | |
| MFCD00063193 | |
| z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu | |
| (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only