missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-glutamic-acid 5-tert-butyl ester, 98%

Manufacturer:  Alfa Aesar™ H6337106

 View more versions of this product

Catalog No. AAH6337106



Specifications

Specifications

N-Benzyloxycarbonyl-D-glutamic-acid 5-tert-butyl ester
51644-83-8
MFCD00077018
GLMODRZPPBZPPB-CYBMUJFWSA-N
(2R)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
6992708
Solid
5g
C17H23NO6
z-d-glu otbu-oh, z-d-glu obut-oh, d-glutamic acid,n-phenylmethoxy carbonyl-, 5-1,1-dimethylethyl ester, n-benzyloxy carbonyl-d-glutamic acid 5-tert-butyl ester, n-alpha-benzyloxycarbonyl-d-glutamic-acid-gamma-t-butyl ester, n-benzyloxycarbonyl-d-glutamic-acid 5-tert-butyl ester, r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
337.372
337.37
98%

Chemical Identifiers

51644-83-8
337.372
GLMODRZPPBZPPB-CYBMUJFWSA-N
6992708
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C17H23NO6
MFCD00077018
z-d-glu otbu-oh, z-d-glu obut-oh, d-glutamic acid,n-phenylmethoxy carbonyl-, 5-1,1-dimethylethyl ester, n-benzyloxy carbonyl-d-glutamic acid 5-tert-butyl ester, n-alpha-benzyloxycarbonyl-d-glutamic-acid-gamma-t-butyl ester, n-benzyloxycarbonyl-d-glutamic-acid 5-tert-butyl ester, r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid
(2R)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
Documents