Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester
CAS 5241-62-3
Formula Weight 357.36
MDL Number MFCD00237336
Beilstein 2570779
Synonym Z-D-Asp(OBzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6532503

  SDS
alfa aesar™
H6532503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6532506

  SDS
alfa aesar™
H6532506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65325MD

  SDS
alfa aesar™
H65325MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 105° to 109°C
Formula Weight 357.36
MDL Number MFCD00237336
Assay 95%
Chemical Name or Material N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester
Physical Form Solid
Beilstein 2570779
Quantity 1g
Storage Note 1 2° to 8°C
CAS 5241-62-3
Synonym Z-D-Asp(OBzl)-OH
Safety and Handling

2° to 8°C