missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H65325MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
105°C to 109°C | |
5241-62-3 | |
MFCD00237336 | |
z-d-asp obzl-oh, z-d-asp-obzl, 2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-benzyloxycarbonyl-d-aspartic acid 4-benzyl ester, 2r-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid non-preferred name | |
OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1 | |
357.36 | |
357.36 | |
250 mg |
N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester | |
C19H19NO6 | |
2570779 | |
VUKCNAATVIWRTF-UHFFFAOYNA-N | |
4-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid | |
47604 | |
95% | |
Solid |
Chemical Identifiers
5241-62-3 | |
357.36 | |
VUKCNAATVIWRTF-UHFFFAOYNA-N | |
47604 | |
OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1 |
C19H19NO6 | |
MFCD00237336 | |
z-d-asp obzl-oh, z-d-asp-obzl, 2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-benzyloxycarbonyl-d-aspartic acid 4-benzyl ester, 2r-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid non-preferred name | |
4-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold