Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-alanylglycine, 95%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-alanylglycine
CAS 34286-66-3
Formula Weight 280.28
MDL Number MFCD00238424
Synonym Z-D-Ala-Gly-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6680203

 
alfa aesar™
H6680203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6680206

 
alfa aesar™
H6680206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66802MD

 
alfa aesar™
H66802MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.28
Chemical Name or Material N-Benzyloxycarbonyl-D-alanylglycine
Physical Form Solid
CAS 34286-66-3
Assay 95%
MDL Number MFCD00238424
Synonym Z-D-Ala-Gly-OH
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C