Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-alaninol, 98%

$62.96 - $837.36

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-alaninol
CAS 61425-27-2
Molecular Formula C11H15NO3
Formula Weight 209.25
MDL Number MFCD00672531
Synonym (2R)-N-Benzyloxycarbonylamino-1-propanol; N-Cbz-D-Alaninol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2237203

  SDS
alfa aesar™
B2237203
1g Each for $62.96

AAB2237206

  SDS
alfa aesar™
B2237206
5g Each for $227.81

AAB2237214

  SDS
alfa aesar™
B2237214
25g Each for $837.36
Description & Specifications

Specifications

Formula Weight 209.25
Chemical Name or Material N-Benzyloxycarbonyl-D-alaninol
Assay 98%
Molecular Formula C11H15NO3
Melting Point 82° to 84°C
MDL Number MFCD00672531
CAS 61425-27-2
Quantity 1g
Synonym (2R)-N-Benzyloxycarbonylamino-1-propanol; N-Cbz-D-Alaninol
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C