Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-D-alanine, 98+%

$39.88 - $39.88

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-D-alanine
CAS 26607-51-2
Molecular Formula C11H13NO4
Formula Weight 223.23
MDL Number MFCD00063126
Beilstein 2056163
Synonym N-Cbz-D-alanine; N-Carbobenzyloxy-D-aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0838203

 
alfa aesar™
L0838203
1g Each for $39.88
Description & Specifications

Specifications

Formula Weight 223.23
Chemical Name or Material N-Benzyloxycarbonyl-D-alanine
MDL Number MFCD00063126
CAS 26607-51-2
Synonym N-Cbz-D-alanine; N-Carbobenzyloxy-D-aniline
Assay ≥98%
Molecular Formula C11H13NO4
Melting Point 83° to 86°C
Beilstein 2056163
Quantity 1g
Optical Rotation +14.5° (c=2 in Acetic Acid)