Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-beta-alanylglycine, 97%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-beta-alanylglycine
CAS 58171-88-3
Formula Weight 280.28
MDL Number MFCD00055930
Beilstein 2008410
Synonym Z-β-Ala-Gly-OH; [3-(Benzyloxycarbonylamino)propanoyl]glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH65424MD

 
alfa aesar™
H65424MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.28
Chemical Name or Material N-Benzyloxycarbonyl-beta-alanylglycine
MDL Number MFCD00055930
CAS 58171-88-3
Physical Form Solid
Assay 97%
Beilstein 2008410
Synonym Z-β-Ala-Gly-OH; [3-(Benzyloxycarbonylamino)propanoyl]glycine
Quantity 250mg