missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester, 98%

$35.32 - $417.92

Chemical Identifiers

CAS 64187-48-0
Molecular Formula C14H17NO5
Molecular Weight (g/mol) 279.292
MDL Number MFCD00055851
InChI Key VVKAGQHUUDRPOI-NEPJUHHUSA-N
Synonym z-hyp-ome, 2s,4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-l-4-hydroxyproline methyl ester, 1-benzyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-trans-4-hydroxy-l-proline methyl ester, trans-n-cbz-4-hydroxy-l-proline methyl ester, l-zhprome, z-l-hyp-ome, z-hydroxy-l-proline methyl ester, 1,2-pyrrolidinedicarboxylicacid, 4-hydroxy-, 2-methyl 1-phenylmethyl ester, 2s,4r
PubChem CID 688411
IUPAC Name 1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES COC(=O)C1CC(CN1C(=O)OCC2=CC=CC=C2)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63993MD View Documents Alfa Aesar™
H63993MD
250mg Each for $35.32
 
AAH6399303 View Documents Alfa Aesar™
H6399303
1g Each for $105.01
 
AAH6399306 View Documents Alfa Aesar™
H6399306
5g Each for $417.92
 
Specifications

Chemical Identifiers

64187-48-0
279.292
VVKAGQHUUDRPOI-NEPJUHHUSA-N
688411
COC(=O)C1CC(CN1C(=O)OCC2=CC=CC=C2)O
C14H17NO5
MFCD00055851
z-hyp-ome, 2s,4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-l-4-hydroxyproline methyl ester, 1-benzyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-trans-4-hydroxy-l-proline methyl ester, trans-n-cbz-4-hydroxy-l-proline methyl ester, l-zhprome, z-l-hyp-ome, z-hydroxy-l-proline methyl ester, 1,2-pyrrolidinedicarboxylicacid, 4-hydroxy-, 2-methyl 1-phenylmethyl ester, 2s,4r
1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate

Specifications

64187-48-0
MFCD00055851
VVKAGQHUUDRPOI-NEPJUHHUSA-N
1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
688411
Liquid
Yellow
N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester
C14H17NO5
z-hyp-ome, 2s,4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-l-4-hydroxyproline methyl ester, 1-benzyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-trans-4-hydroxy-l-proline methyl ester, trans-n-cbz-4-hydroxy-l-proline methyl ester, l-zhprome, z-l-hyp-ome, z-hydroxy-l-proline methyl ester, 1,2-pyrrolidinedicarboxylicacid, 4-hydroxy-, 2-methyl 1-phenylmethyl ester, 2s,4r
COC(=O)C1CC(CN1C(=O)OCC2=CC=CC=C2)O
279.292
279.29
98%
250mg
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C