missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester, 98%

Manufacturer:  Alfa Aesar™ H63993MD

 View more versions of this product

Catalog No. AAH63993MD



Specifications

Specifications

N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester
C14H17NO5
12-pyrrolidinedicarboxylicacid 4-hydroxy- 2-methyl 1-phenylmethyl ester 2s4r, 2s 4r-4-hydroxy-pyrrolidine-12-dicarboxylic acid 1-benzyl ester 2-methyl ester, 2s4r-4-hydroxy-pyrrolidine-12-dicarboxylic acid 1-benzyl ester 2-methyl ester, methyl 2s4r-1-benzyloxycarbonyl-4-hydroxypyrrolidine-2-carboxylate, 1-o-benzyl 2-o-methyl 2s4r-4-hydroxypyrrolidine-12-dicarboxylate, 2s4r-1-benzyloxycarbonyl-4-hydroxy-2-methoxycarbonylpyrrolidine, 2s4r-1-benzyloxycarbonyl-2-methoxycarbonyl-4-hydroxypyrrolidine, 1-benzyl 2-methyl 2s4r-4-hydroxypyrrolidine-12-dicarboxylate, 2s4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-12-dicarboxylate
COC(=O)C1CC(CN1C(=O)OCC2=CC=CC=C2)O
279.292
279.29
98%
250mg
64187-48-0
MFCD00055851
VVKAGQHUUDRPOI-NEPJUHHUSA-N
1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
688411
Liquid
Yellow

Chemical Identifiers

64187-48-0
279.292
VVKAGQHUUDRPOI-NEPJUHHUSA-N
688411
COC(=O)C1CC(CN1C(=O)OCC2=CC=CC=C2)O
C14H17NO5
MFCD00055851
12-pyrrolidinedicarboxylicacid 4-hydroxy- 2-methyl 1-phenylmethyl ester 2s4r, 2s 4r-4-hydroxy-pyrrolidine-12-dicarboxylic acid 1-benzyl ester 2-methyl ester, 2s4r-4-hydroxy-pyrrolidine-12-dicarboxylic acid 1-benzyl ester 2-methyl ester, methyl 2s4r-1-benzyloxycarbonyl-4-hydroxypyrrolidine-2-carboxylate, 1-o-benzyl 2-o-methyl 2s4r-4-hydroxypyrrolidine-12-dicarboxylate, 2s4r-1-benzyloxycarbonyl-4-hydroxy-2-methoxycarbonylpyrrolidine, 2s4r-1-benzyloxycarbonyl-2-methoxycarbonyl-4-hydroxypyrrolidine, 1-benzyl 2-methyl 2s4r-4-hydroxypyrrolidine-12-dicarboxylate, 2s4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-12-dicarboxylate
1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents