missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-4-hydroxy-L-proline, 99%

$84.96 - $294.54

Chemical Identifiers

CAS 13504-85-3
Molecular Formula C13H15NO5
Molecular Weight (g/mol) 265.265
MDL Number MFCD00037329
InChI Key WWVCWLBEARZMAH-DTIOYNMSSA-N
Synonym 2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, z-l-4-hydroxyproline, n-cbz-4-hydroxyproline, n-cbz-4-hydroxy-l-proline, n-benzyloxycarbonyl-4-hydroxyproline, n-carbobenzyloxy-4-hydroxy-l-proline, 4-hydroxy-n-benzyloxycarbonyl-l-proline, 1-carbobenzyloxy-4-hydroxy-l-proline, s-1-cbz-4-hydroxy-2-pyrrolidinecarboxylic acid
PubChem CID 12731799
IUPAC Name (2S)-4-hydroxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1C(CN(C1C(=O)O)C(=O)OCC2=CC=CC=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5988206 View Documents Alfa Aesar™
H5988206
5g Each for $84.96
 
AAH5988214 View Documents Alfa Aesar™
H5988214
25g Each for $294.54
 
Description

Description

Applications
A competitive inhibitor of porcine kidney prolidase.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

13504-85-3
265.265
WWVCWLBEARZMAH-DTIOYNMSSA-N
12731799
C1C(CN(C1C(=O)O)C(=O)OCC2=CC=CC=C2)O
C13H15NO5
MFCD00037329
2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, z-l-4-hydroxyproline, n-cbz-4-hydroxyproline, n-cbz-4-hydroxy-l-proline, n-benzyloxycarbonyl-4-hydroxyproline, n-carbobenzyloxy-4-hydroxy-l-proline, 4-hydroxy-n-benzyloxycarbonyl-l-proline, 1-carbobenzyloxy-4-hydroxy-l-proline, s-1-cbz-4-hydroxy-2-pyrrolidinecarboxylic acid
(2S)-4-hydroxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid

Specifications

−54° (c=2 in Ethanol)
5g
C13H15NO5
90295
WWVCWLBEARZMAH-DTIOYNMSSA-N
(2S)-4-hydroxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
12731799
99%
104°C to 107°C
13504-85-3
MFCD00037329
2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, z-l-4-hydroxyproline, n-cbz-4-hydroxyproline, n-cbz-4-hydroxy-l-proline, n-benzyloxycarbonyl-4-hydroxyproline, n-carbobenzyloxy-4-hydroxy-l-proline, 4-hydroxy-n-benzyloxycarbonyl-l-proline, 1-carbobenzyloxy-4-hydroxy-l-proline, s-1-cbz-4-hydroxy-2-pyrrolidinecarboxylic acid
C1C(CN(C1C(=O)O)C(=O)OCC2=CC=CC=C2)O
265.265
265.26
N-Benzyloxycarbonyl-4-hydroxy-L-proline
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C