missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyloxycarbonyl-3-(Fmoc-amino)-L-alanine, 95%, Thermo Scientific™

$59.89 - $629.03

Chemical Identifiers

CAS 142855-80-9
MDL Number MFCD00274016
Synonym Z-Dap(Fmoc)-OH
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66859MD
View Documents Promotion Details
Thermo Scientific Chemicals
H66859MD
250 mg
Each for $59.89
 
AAH6685903
View Documents Promotion Details
Thermo Scientific Chemicals
H6685903
1 g
Each for $169.75
 
AAH6685906
View Documents Promotion Details
Thermo Scientific Chemicals
H6685906
5 g
Each for $629.03
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

142855-80-9
MFCD00274016
Z-Dap(Fmoc)-OH

Specifications

142855-80-9
MFCD00274016
Z-Dap(Fmoc)-OH
95%
Powder
White
6356947
460.49
250 mg
N-Benzyloxycarbonyl-3-(Fmoc-amino)-L-alanine
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'