missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-1H-pyrazole-1-carboxamidine, 98+%

Manufacturer:  Alfa Aesar™ H5478206

 View more versions of this product

Catalog No. AAH5478206



Specifications

Specifications

N-Benzyloxycarbonyl-1H-pyrazole-1-carboxamidine
5g
C12H12N4O2
benzyl imino 1h-pyrazol-1-yl methyl carbamate
C1=CC=C(C=C1)COC(=O)N=C(N)N2C=CC=N2
244.254
244.25
106°C to 110°C
152120-62-2
MFCD00274657
BXUMISWXFKCEIA-UHFFFAOYSA-N
benzyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
11857377
≥98%

Chemical Identifiers

152120-62-2
244.254
BXUMISWXFKCEIA-UHFFFAOYSA-N
11857377
C1=CC=C(C=C1)COC(=O)N=C(N)N2C=CC=N2
C12H12N4O2
MFCD00274657
benzyl imino 1h-pyrazol-1-yl methyl carbamate
benzyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents