missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-1H-pyrazole-1-carboxamidine, 98+%

Manufacturer:  Alfa Aesar™ H5478203

 View more versions of this product

Catalog No. AAH5478203



Specifications

Specifications

N-Benzyloxycarbonyl-1H-pyrazole-1-carboxamidine
1g
C12H12N4O2
benzyl imino 1h-pyrazol-1-yl methyl carbamate
C1=CC=C(C=C1)COC(=O)N=C(N)N2C=CC=N2
244.254
244.25
106°C to 110°C
152120-62-2
MFCD00274657
BXUMISWXFKCEIA-UHFFFAOYSA-N
benzyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
11857377
≥98%

Chemical Identifiers

152120-62-2
244.254
BXUMISWXFKCEIA-UHFFFAOYSA-N
11857377
C1=CC=C(C=C1)COC(=O)N=C(N)N2C=CC=N2
C12H12N4O2
MFCD00274657
benzyl imino 1h-pyrazol-1-yl methyl carbamate
benzyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents