missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 427070050

 View more versions of this product

Catalog No. AC427070050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-Benzyloxycarbonyl-1, 3-propanediamine hydrochloride
97.5
Authentic
Glass bottle
MFCD00270153
n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
[H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1
244.72
244.72
17400-34-9
100.0
98%
C11H17ClN2O2
5g
XKMBTMXQMDLSRB-UHFFFAOYSA-N
benzyl N-(3-aminopropyl)carbamate;hydrochloride
13196227
98%

Chemical Identifiers

17400-34-9
244.72
XKMBTMXQMDLSRB-UHFFFAOYSA-N
13196227
[H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1
C11H17ClN2O2
MFCD00270153
n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
benzyl N-(3-aminopropyl)carbamate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only