Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride
CAS 17400-34-9
Formula Weight 244.72
MDL Number MFCD00270153
Beilstein 4725495
Synonym Benzyl-(3-aminopropyl)carbamate hydrochloride; Z-1,3-diaminopropane.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6310303

  SDS
alfa aesar™
H6310303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6310306

  SDS
alfa aesar™
H6310306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.72
Melting Point 185° to 189°C
Chemical Name or Material N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride
Physical Form Crystalline Solid
Assay 97%
MDL Number MFCD00270153
CAS 17400-34-9
Beilstein 4725495
Color White
Quantity 1g
Synonym Benzyl-(3-aminopropyl)carbamate hydrochloride; Z-1,3-diaminopropane.HCl
Sensitivity Hygroscopic