missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H6310306

 View more versions of this product

Catalog No. AAH6310306



Specifications

Specifications

N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride
White
5g
C11H17ClN2O2
4725495
XKMBTMXQMDLSRB-UHFFFAOYSA-N
benzyl N-(3-aminopropyl)carbamate;hydrochloride
13196227
Crystalline Solid
Hygroscopic
185°C to 189°C
17400-34-9
MFCD00270153
n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
C1=CC=C(C=C1)COC(=O)NCCCN.Cl
244.719
244.72
97%

Chemical Identifiers

17400-34-9
244.719
XKMBTMXQMDLSRB-UHFFFAOYSA-N
13196227
C1=CC=C(C=C1)COC(=O)NCCCN.Cl
C11H17ClN2O2
MFCD00270153
n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
benzyl N-(3-aminopropyl)carbamate;hydrochloride
SDS Documents