Please login to your online account to display your discounted pricing

N-Benzylnaltrindole hydrochloride, Tocris


δ2 selective non-peptide antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS075410

 View more versions of this product

Catalog No. 075410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 541.09
Chemical Name or Material N-Benzylnaltrindole hydrochloride
Assay Percent Range >98%
Molecular Formula C33H32N2O3·HCl
CAS 1206487-81-1
Synonym 1'-Benzyl-17-(cyclopropylmethyl)-6,7-didehydro-4,5a-epoxy-3,14-dihydroxyindolo[2',3':6,7]morphinan hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material N-Benzylnaltrindole hydrochloride
Assay Percent Range >98%
Molecular Formula C33H32N2O3·HCl
Formula Weight 541.09
Synonym 1'-Benzyl-17-(cyclopropylmethyl)-6,7-didehydro-4,5a-epoxy-3,14-dihydroxyindolo[2',3':6,7]morphinan hydrochloride
Accessories

Store at Room Temperature