Please login to your online account to display your discounted pricing

N-Benzylnaltrindole hydrochloride, Tocris

δ2 selective non-peptide antagonist

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material N-Benzylnaltrindole hydrochloride
Assay Percent Range >98%
Molecular Formula C33H32N2O3·HCl
Formula Weight 541.09
Synonym 1'-Benzyl-17-(cyclopropylmethyl)-6,7-didehydro-4,5a-epoxy-3,14-dihydroxyindolo[2',3':6,7]morphinan hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

075410

 
r & d systems
RDS075410
10mg Each for $208.25

075450

 
r & d systems
RDS075450
50mg Each for $867.95
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 541.09
Chemical Name or Material N-Benzylnaltrindole hydrochloride
Assay Percent Range >98%
Molecular Formula C33H32N2O3·HCl
CAS 1206487-81-1
Synonym 1'-Benzyl-17-(cyclopropylmethyl)-6,7-didehydro-4,5a-epoxy-3,14-dihydroxyindolo[2',3':6,7]morphinan hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature