Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Benzylmaleimide, 99%

Chemical Identifiers

Chemical Name or Material N-Benzylmaleimide
CAS 1631-26-1
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00014540
UN Number UN1759
Beilstein 135801
Synonym 1-Benzyl-1H-pyrrole-2,5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1355106

 
alfa aesar™
A1355106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1355122

 
alfa aesar™
A1355122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.2
UN Number UN1759
Quantity 5g
Chemical Name or Material N-Benzylmaleimide
Assay 99%
Molecular Formula C11H9NO2
Melting Point 67° to 70°C
MDL Number MFCD00014540
CAS 1631-26-1
Beilstein 135801
Synonym 1-Benzyl-1H-pyrrole-2,5-dione
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Applications
N-Benzylmaleimide is used as plastic additive, Pharmaceutical Intermediates. It is an important raw material intermediate used in organic Synthesis, Pharmaceuticals, Agrochemicals Dyestuff.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.