Please login to your online account to display your discounted pricing

N-Benzylmaleimide, 98%, ACROS Organics™

$23.88 - $23.88

Chemical Identifiers

Chemical Name or Material N-Benzylmaleimide
CAS 1631-26-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H9NO2
Formula Weight 187.2
MDL Number MFCD00014540
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334190010

  SDS
thermo fisher scientific
AC334190010
Glass bottle 1g Each for $23.88
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder or Needles
Formula Weight 187.2
Quantity 1g
Color Off-white to light yellow
Melting Point 68° to 71°C
CAS Max % 100.0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
CAS 1631-26-1
Molecular Formula C11H9NO2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00014540
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material N-Benzylmaleimide
Assay Percent Range 98%
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.